EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Anthrone-derived NHPI analogues as catalysts in reactions using oxygen as an oxidant.
PMID 18972037
Abstract
An enantioselective synthesis of anthrone-derived NHPI analogues has been developed. One of these analogues, in combination with Co salts, was employed to catalyse the aerobic oxidation of benzylic compounds and diols. Exploratory studies using a racemic version of the catalyst were also conducted. Radical addition of dioxolanes or alcohols to activated alkenes with molecular oxygen as the terminal oxidant was also shown to be catalysed with NHPI analogues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
319899
Anthrone, ACS reagent, 97%
C14H10O
H53704
N-Hydroxyphthalimide, 97%
C8H5NO3