EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Ligand specificity of constitutive androstane receptor as probed by induced-fit docking and mutagenesis.
PMID 18973326
Abstract
Constitutive androstane receptor (CAR, NR1I3) belongs to the nuclear receptor family of transcription factors and acts as a chemical sensor of drugs and endogenous compounds. The ligand-binding preferences of CAR are diverse, and more importantly, there are significant species differences in ligand specificity. Here, we show that while certain residues are critical for the basal activity of mouse CAR (mCAR) and/or affect the binding of all tested ligands, mutation of some ligand-binding pocket (LBP) residues (e.g., F171 and Y336) paradoxically decreased the activity of a specific ligand while increasing that of others. Comparisons to previously reported human CAR (hCAR) residues indicated that the function of key CAR residues (e.g., N175, L253) is dramatically different between species. The docking results provide some mechanistic rationale for the ability of 17alpha-ethinyl-3,17beta-estradiol (EE2) to both activate mCAR and repress hCAR.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7883
5α-Androst-16-en-3α-ol
C19H30O
36161
Methoxychlor, PESTANAL®, analytical standard
C16H15Cl3O2