EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Isolation and in vitro binding of mating type plus fertilization tubules from Chlamydomonas.
PMID 18979246
Abstract
During fertilization in Chlamydomonas, adhesion and fusion of gametes occur at the tip of specialized regions of the plasma membrane, known as mating structures. The mating type minus (mt[-]) structure is a slightly raised dome-shaped region located at the apical end of the cell body. In contrast, the activated mating type plus (mt[+]) structure is an actin-filled, microvillouslike organelle. Interestingly, a similar type of "fusion organelle" is conserved across diverse groups. Chlamydomonas provides an ideal model system for studying the process of gametic cell fusion in that it is amenable to genetic manipulations as well as cell and molecular biological approaches. Moreover, the ease of culturing Chlamydomonas combined with the ability to isolate the mt(+) fertilization tubule and the development of in vitro assays for adhesion makes it an ideal system for biochemical studies focused on dissecting the molecular mechanisms that underlie the complex process of gametic cell fusion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8157
Phallacidin from Amanita phalloides, ≥85%
C37H50N8O13S