EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
Biobehavioral influences on matrix metalloproteinase expression in ovarian carcinoma.
PMID 18980978
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB13458 Anti-MMP-9 Antibody