EMAIL THIS PAGE TO A FRIEND
Canadian journal of ophthalmology. Journal canadien d'ophtalmologie
Bilateral infiltrative orbital disease: is it chemically induced?
PMID 18982043
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72568
Nitrilotri(methylphosphonic acid), ≥97.0% (T)
C3H12NO9P3