EMAIL THIS PAGE TO A FRIEND
Journal of thrombosis and haemostasis : JTH
Phosphatidylethanolamine at the endothelial surface of aortic flow dividers.
PMID 18983507
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3168 Duramycin from Streptoverticillium cinnamoneus, ≥90.0%
C89H125N23O25S3