EMAIL THIS PAGE TO A FRIEND
Chemistry and physics of lipids
Free radical fragmentation of cardiolipin by cytochrome c.
PMID 18983994
Abstract
The effect of cytochrome c (cyt c) on degradation of cardiolipin in its polar part was investigated in cardiolipin/phosphatidylcholine (CL/PC) liposomes incubated with cyt c/H(2)O(2)/and (or) ascorbate by high-performance thin layer chromatography and MALDI-TOF mass spectrometry. It has been shown that phosphatidic acid (PA) and phosphatidylhydroxyacetone (PHA) were formed in the system under conditions where hydrogen peroxide favours a release of heme iron from cyt c. The formation of PA and PHA occurs via an OH-induced fragmentation taking place in the polar moiety of cardiolipin. Formation of fragmentation products correlated with the loss of CL in CL/PC liposomes incubated with cyt c/H(2)O(2)/ascorbate or with Cu(2+)/H(2)O(2)/ascorbate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C3131
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis