Prostaglandins & other lipid mediators
Hypoxia suppresses KV1.5 channel expression through endogenous 15-HETE in rat pulmonary artery.
PMID 18984061
Hypoxia initiated pulmonary vasoconstriction is due to the inhibition of voltage-gated K(+) (K(V)) channels. But the mechanism is unclear. We have evidence that hypoxia activates 15-lipoxygenase (15-LOX) in distal pulmonary arteries and increases the formation of 15-hydroxyeicosatetraenoate (15-HETE). 15-HETE-induced pulmonary artery constriction to be through the inhibition of K(V) channels (K(V)1.5, K(V)2.1 and K(V)3.4). However, no direct link among hypoxia, 15-HETE and inhibition of K(V) subtypes is established. Therefore, we investigated whether 15-LOX/15-HETE pathway contributes to the hypoxia-induced down-regulation of K(V) channels. As K(V)1.5 channel is O(2)-sensitive, it was chosen in the initial study. We found that inhibition of 15-LOX suppressed the response of hypoxic pulmonary artery rings to phenylephrine. The expressions of K(V)1.5 channel mRNA and protein was robustly up-regulated in cultured PASMC and pulmonary artery after blocking of 15-LOX by lipoxygenase inhibitors in hypoxia. The 15-LOX blockade also partly rescued the voltage-gated K(+) current (I(K(V))). 15-HETE contributes to the down-regulation of K(V)1.5 channel, inhibition of I(K(V)) and increase of native pulmonary artery tension after hypoxia. Hypoxia inhibits K(V)1.5 channel through 15-LOX/15-HETE pathway.