EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
Identification of phosphorylation sites in the repetitive carboxyl-terminal domain of the mouse RNA polymerase II largest subunit
PMID 1899239