EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Inconspicamide, new N-acylated serinol from the marine sponge Stelletta inconspicua.
PMID 18997401
Abstract
A new N-acylated serinol, inconspicamide (1), was isolated from the marine sponge, Stelletta inconspicua, together with a glyceryl ether (2). Their structures were determined on the basis of spectroscopic data and the modified Mosher analysis. They exhibited moderate cytotoxic activity against HeLa human cervical cancer cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357898
Serinol, 98%
C3H9NO2