EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Directed epoxidation of cyclohexa-1,4-dienes-stereoselective formation of up to six contiguous stereogenic centres.
PMID 19005603
Abstract
Epoxidation/cyclisation of cyclohexa-1,4-dienes containing pendant hydroxyl groups provides stereocontrolled access to highly-functionalised reduced benzol[b]furan derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
28910
1,4-Cyclohexadiene, purum, ≥97.0% (GC)
C6H8
125415
1,4-Cyclohexadiene, 97%
C6H8