EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
From glycosylation disorders back to glycosylation: what have we learned?
PMID 19007883
Abstract
Diseases of glycosylation have long remained confined to the rare hematological disorders, the Tn-syndrome and paroxysmal nocturnal hemoglobinuria. This rarity was often interpreted as a sign that defects of glycosylation are either lethal, or remain asymptomatic because of the large redundancy found in glycosylation pathways. The description of multiple glycosylation disorders over the last years has definitively settled the issue and demonstrated the broad range of biological processes relying on proper glycosylation. However, beyond establishing the developmental and physiological roles of glycosylation how did glycosylation disorders provided new insights to the field of glycobiology?
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8271
Cytidine-5′-monophospho-N-acetylneuraminic acid sodium salt, ≥85% (HPLC)
C20H30N4NaO16P