EMAIL THIS PAGE TO A FRIEND
Neuropharmacology
Antinociceptive effects of the marine snail peptides conantokin-G and conotoxin MVIIA alone and in combination in rat models of pain.
PMID 19010337
Abstract
There are a number of neurologically active ion channel blocking peptides derived from cone snail venom, such as conantokin-G and omega-conotoxin MVIIA. Conantokin-G inhibits NMDA receptors containing the NR2B subunit whereas omega-conotoxin MVIIA blocks N-type Ca(2+) channels. Separately, these peptides induce antinociceptive effects in pre-clinical pain models following intrathecal injection. In the current study, the efficacies of these peptides were determined separately and in combination by intrathecal injection into rats with a spinal nerve ligation, in rats with a spinal cord compression injury and in the formalin test. Separately, both conantokin-G and omega-conotoxin MVIIA dose-dependently attenuated nociceptive responses in all of these models. However, at high antinociceptive doses for both formalin and nerve injury models, omega-conotoxin MVIIA evoked untoward side effects. Using isobolographic analysis, the combination of sub-antinociceptive doses of peptides demonstrated additive antinociception in rats with a nerve ligation and in the formalin test, without apparent adverse side effects. In a model of neuropathic spinal cord injury pain, which is clinically difficult to treat, the combination of conantokin-G and omega-conotoxin MVIIA resulted in robust synergistic antinociception. These data suggest that a combination of these peptides may be analgesic across diverse clinical pains with limited untoward side effects, and particularly potent for reducing spinal cord injury pain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4311
Conantokin G, ≥75% (HPLC)
C88H139N26O44