EMAIL THIS PAGE TO A FRIEND
Advances in chromatography
Liquid chromatographic separations of basic compounds.
PMID 19013951
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
211982
Solvent Green 3, Dye content 95 %
C28H22N2O2