EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Cross-coupling of aryl/alkenyl pivalates with organozinc reagents through nickel-catalyzed C-O bond activation under mild reaction conditions.
PMID 19016280
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T71803
Pivalic acid, 99%
C5H10O2