EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Ligand oxidation of a deprotonated bis(picolyl)amine Ir(I)(cod) complex.
PMID 19016555
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A65409
3-Picolylamine, ≥99%
C6H8N2