EMAIL THIS PAGE TO A FRIEND
The European journal of neuroscience
Role of spinal 5-HT2 receptor subtypes in quipazine-induced hindlimb movements after a low-thoracic spinal cord transection.
PMID 19019202
Abstract
A role of serotonin receptors (5-HTRs) in spinal rhythmogenesis has been proposed several years ago based mainly upon data showing that bath-applied 5-HT could elicit locomotor-like rhythms in in vitro isolated spinal cord preparations. Such a role was partially confirmed in vivo after revealing that systemically administered 5-HTR(2) agonists, such as quipazine, could induce some locomotor-like movements (LM) in completely spinal cord-transected (Tx) rodents. However, given the limited binding selectivity of currently available 5-HTR(2) agonists, it has remained difficult to determine clearly if one receptor subtype is specifically associated with LM induction. In situ hybridization, data using tissues from L1-L2 spinal cord segments, where critical locomotor network elements have been identified in mice, revealed greater 5-HTR(2A) mRNA levels in low-thoracic Tx than non-Tx animals. This expression level remained elevated for several days, specifically in the lateral intermediate zone, where peak values were detected at 1 week post-Tx and returned to normal at 3 weeks post-Tx. Behavioral and kinematic analyses revealed quipazine-induced LM in 1-week Tx mice either non-pretreated or pretreated with selective 5-HTR(2B) and/or 5-HTR(2C) antagonists. In contrast, LM completely failed to be induced by quipazine in animals pretreated with selective 5-HTR(2A) antagonists. Altogether, these results provide strong evidence suggesting that 5-HTR(2A) are specifically associated with spinal locomotor network activation and LM generation induced by quipazine in Tx animals. These findings may contribute to design drug treatments aimed at promoting locomotor function recovery in chronic spinal cord-injured patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Q1004
Quipazine maleate salt
C13H15N3 · C4H4O4