EMAIL THIS PAGE TO A FRIEND
Water environment research : a research publication of the Water Environment Federation
Solid-phase extraction and preconcentration of copper in mineral waters with 4-(2-pyridyl-azo) resorcinol-loaded amberlite XAD-7 and flame atomic absorption spectrometry.
PMID 19024725
Abstract
A simple, sensitive, accurate, and selective method for determination of ultratrace levels of copper is modified. The method is based on preconcentration of copper on the 4-(2-pyridyl-azo) resorcinol-loaded amberlite XAD-7 (non-ionic copoly[styrene-acrylic compound]) at pH 5.0 to 6.5 for contact time as low as 45 minutes. The adsorbed copper was eluted with concentrated nitric acid and measured by flame atomic absorption spectrometry. Recoveries of up to 90% were achieved. The optimized preconcentration method was applied to copper determination in various water samples. The detection limit was found to be 0.09 ng mL(-1). The relative standard deviation was found to be 9% using 300 mL of 5.0 ng mL(-1) for 10 replicate preconcentration procedures. Copper concentrations in the studied water samples were found to be in the ranges of 0.4 to 18.0 ng mL(-1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
323209
4-(2-Pyridylazo)resorcinol, 96%
C11H9N3O2