EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Three-dimensional structure of recombinant human interferon-gamma.
PMID 1902591
Abstract
The x-ray crystal structure of recombinant human interferon-gamma has been determined with the use of multiple-isomorphous-replacement techniques. Interferon-gamma, which is dimeric in solution, crystallizes with two dimers related by a noncrystallographic twofold axis in the asymmetric unit. The protein is primarily alpha helical, with six helices in each subunit that comprise approximately 62 percent of the structure; there is no beta sheet. The dimeric structure of human interferon-gamma is stabilized by the intertwining of helices across the subunit interface with multiple intersubunit interactions.