EMAIL THIS PAGE TO A FRIEND
Free Radical Biology & Medicine
HPLC study of oxidation products of hydroethidine in chemical and biological systems: ramifications in superoxide measurements.
PMID 19026738