EMAIL THIS PAGE TO A FRIEND
Journal of Chemical Neuroanatomy
Triple fluorescence labelling of neuronal, glial and vascular markers revealing pathological alterations in various animal models.
PMID 19028564
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB377
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse