EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Molecular evidence for spread of two major methicillin-resistant Staphylococcus aureus clones with a unique geographic distribution in Chinese hospitals.
PMID 19029328
Abstract
Methicillin (meticillin)-resistant Staphylococcus aureus (MRSA) is a serious problem worldwide. To investigate the molecular epidemiology of MRSA isolates in China, a total of 702 MRSA isolates collected from 18 teaching hospitals in 14 cities between 2005 and 2006 were characterized by antibiogram analysis, pulsed-field gel electrophoresis (PFGE), staphylococcal cassette chromosome mec (SCCmec) typing, and spa typing; and 102 isolates were selected for multilocus sequence typing (MLST). Overall, SCCmec type III was the most popular type and was found in 541 isolates (77.1%), followed by SCCmec type II (109/702; 15.5%). Twenty-four PFGE types were obtained among 395 isolates collected in 2005, and 18 spa types were obtained among 702 isolates. spa type t030, which corresponded to PFEG types A to E, constituted 52.0% (365/702) of all isolates, and isolates of this type were present in all 14 cities; spa type t037, which corresponded to PFGE types F and G, accounted for 25.5% (179/702) of all isolates, and isolates of this type were identified in 12 cities. The two spa genotypes belonged to sequence type 239 (ST239) and carried SCCmec type III. spa type t002, which included isolates of PFGE types L to T, made up 16.0% (112/702) of the isolates that belonged to ST5 and SCCmec type II, and isolates of this type were distributed in 12 cities. The distribution of spa types varied among the regions. spa type t002 was the most common in Dalian (53.4%) and Shenyang (44.4%); spa type t037 was predominant in Shanghai (74.8%), whereas spa type t030 was the most common in the other cities. Two isolates from Guangzhou that harbored SCCmec type IVa with ST59 and ST88 were identified as community-associated MRSA. The prevalence of the Panton-Valentine leukocidin gene was 2.3%. The data documented two major epidemic MRSA clones, ST239-MRSA-SCCmec type III and ST5-MRSA-SCCmec type II, with unique geographic distributions across China.