EMAIL THIS PAGE TO A FRIEND
Australian endodontic journal : the journal of the Australian Society of Endodontology Inc
Evaluation of the temperature of different refrigerant sprays used as a pulpal test.
PMID 19032640
Abstract
The temperature of different refrigerant sprays (Endo-Ice, Endo-Frost, Coolermatic and Sprayon Contact and Tuner Cleaner) used as pulpal tests were evaluated in vitro. A thermocouple placed inside the pulp chamber of a maxillary central incisor was used to register the temperature changes when the refrigerant sprays were applied with a cotton swab, for 10 s. Results indicate that Endo-Ice and Endo-Frost presented the lowest temperatures among the refrigerant sprays tested. Temperatures measured inside the pulp chamber, however, were statistically similar in all groups.