EMAIL THIS PAGE TO A FRIEND
International journal of cancer
CXCL8/IL-8 and CXCL12/SDF-1alpha co-operatively promote invasiveness and angiogenesis in pancreatic cancer.
PMID 19035451
Abstract
CXC-chemokines are involved in the chemotaxis of neutrophils, lymphocytes and monocytes. However, role of these chemokines in tumorigenesis, especially with regard to interaction between tumor and its microenvironment, has not been clearly elucidated. The purpose of this study was to analyze the co-operative role of CXCL8 and CXCL12 in the tumor-stromal interaction in pancreatic cancer (PaCa). Using enzyme-linked immunosorbent assay (ELISA) and reverse transcription polymerase chain reaction (RT-PCR), we initially confirmed the expression of ligands and receptors, respectively, of CXC-chemokines in PaCa and stromal cells. We examined the co-operative role of CXCL8 and CXCL12 in proliferation/invasion of PaCa and human umbilical vein endothelial cells (HUVECs), and in HUVEC tube-formations through tumor-stromal interaction by MTS, Matrigel invasion, and angiogenesis assays, respectively. We detected expression of CXCR4, but not CXCR2, in all PaCa cells and fibroblasts. PaCa cells secreted CXCL8, and fibroblast cells secreted CXCL12. CXCL8 production in PaCa was significantly enhanced by CXCL12, and CXCL12 production in fibroblasts was significantly enhanced by co-culturing with PaCa. CXCL8 enhanced proliferation/invasion of HUVECs but did not promote proliferation/invasion of PaCa. Both recombinant and PaCa-derived CXCL8 enhanced tube formation of HUVECs that were co-cultured with fibroblast cells. CXCL12 enhanced the proliferation/invasion of HUVECs and the invasion of PaCa cells but had no effect on tube formation of HUVEC. We showed that PaCa-derived CXCL8 and fibroblast-derived CXCL12 cooperatively induced angiogenesis in vitro by promoting HUVEC proliferation, invasion, and tube formation. Thus, corresponding receptors CXCR2 and CXCR4 are potential antiangiogenic and antimetastatic therapeutic targets in PaCa.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R7509 RPMI-1640 Medium, Modified, with sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
R7388 RPMI-1640 Medium, Modified, with 20 mM HEPES and L-glutamine, without sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
R8755 RPMI-1640 Medium, Modified, with L-glutamine, without phenol red and sodium bicarbonate, powder, suitable for cell culture
R4130 RPMI-1640 Medium, HEPES Modification, with L-glutamine and 25mM HEPES, without sodium bicarbonate, powder, suitable for cell culture
R7755 RPMI-1640 Medium, AutoMod, without L-glutamine and sodium bicarbonate, powder, suitable for cell culture
R1383 RPMI-1640 Medium, With L-glutamine, without glucose and sodium bicarbonate, powder, suitable for cell culture
R1780 RPMI-1640 Medium, With L-glutamine and sodium bicarbonate. Without arginine, leucine, lysine, and phenol red, liquid, sterile-filtered, suitable for cell culture, designed for isotope labeling for cell culture applications
R8005 RPMI-1640 Medium, Hybri-Max, Modified, with L-glutamine, 4500 mg/L glucose and 15mM HEPES, without sodium bicarbonate, powder, suitable for hybridoma
51536C RPMI-1640 Medium, with 2.05 mM L-glutamine, with 25mM HEPES, liquid, sterile-filtered, suitable for cell culture
R1145 RPMI-1640 Medium, 10 ×, Without L-glutamine, folic acid and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
R7513 RPMI-1640 Medium, Modified, with sodium bicarbonate, without methionine, cystine and L-glutamine, liquid, sterile-filtered, suitable for cell culture
R7638 RPMI-1640 Medium, Dutch Modification, with sodium bicarbonate and 20mM HEPES, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
R2405 StableCell RPMI-1640, With stable glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture