EMAIL THIS PAGE TO A FRIEND
The Science of the total environment
Microbial dehalogenation of 4,4'-dichlorobiphenyl under anaerobic conditions.
PMID 1903889
Abstract
Anaerobic cultures containing 4,4'-dichlorobiphenyl (4,4'CB) were inoculated with various environmental samples. The degradation of the substrate was followed by gas chromatographic analysis. Three of the cultures tested showed an ability to dehalogenate the substrate, as judged by the presence of 4-chlorobiphenyl and biphenyl in those media inoculated with them.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
48260-U 4,4′-Dichlorobiphenyl solution, certified reference material