EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Molecular cloning and sequence analysis of the sisomicin biosynthetic gene cluster from Micromonospora inyoensis.
PMID 19039528
Abstract
Micromonospora inyoensis produces sisomicin (Sm), an aminoglycoside antibiotic. The gene cluster of sisomicin biosynthesis spanning ca. 47 kb consists of 37 ORFs encoding various proteins for sisomicin biosynthesis, regulation, resistance and transport. The comparative genetic studies on the biosynthetic genes of sisomicin and gentamicin (Gm) reveal a similar biosynthetic route and provide a framework for the future biosynthetic studies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S7796
Sisomicin sulfate salt, ≥80% (TLC)
C19H37N5O7 · 2.5H2O4S