EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Naturforschung. C, Journal of biosciences
Flow cytometry and phytochemical analysis of a sunflower cell suspension culture in a 5-L bioreactor.
PMID 19040110
Abstract
A cell suspension culture of sunflower (Helianthus annuus), a producer of immunologically active polysaccharides, was cultivated in a 5-L stirred tank bioreactor, operated in batch mode. After some changes in the internal bioreactor design a stable growth of Helianthus cells was achieved and the accumulated biomass reached 15.2 g/L (only approximately 5% lower compared to the accumulated biomass in shake-flasks). Flow cytometry used for measuring the cell cycle parameters of suspended Helianthus cells did not reveal significant differences between shake-flasks and bioreactor cultivation modes. For both cultivation methods significant enhancement of the percentage of S-phase cells was observed at the beginning of the cultivation process. Concerning the metabolite production the maximum in exopolysaccharides was reached at day 9 of the cultivation period (1.9 g/L), while the highest amounts of alpha-tocopherol were accumulated at the beginning of the cultivation process (day 2 of the cultivation). These finding were related to the respective stress levels caused by the inoculation procedure. The kinetic parameters of growth and polysaccharide production as well as the time course of carbon source utilization were monitored and discussed.