EMAIL THIS PAGE TO A FRIEND
Cell calcium
TRPM4 regulates migration of mast cells in mice.
PMID 19046767
Abstract
We demonstrate here that the transient receptor potential melastatin subfamily channel, TRPM4, controls migration of bone marrow-derived mast cells (BMMCs), triggered by dinitrophenylated human serum albumin (DNP-HSA) or stem cell factor (SCF). Wild-type BMMCs migrate after stimulation with DNP-HSA or SCF whereas both stimuli do not induce migration in BMMCs derived from TRPM4 knockout mice (trpm4(-/-)). Mast cell migration is a Ca(2+)-dependent process, and TRPM4 likely controls this process by setting the intracellular Ca(2+) level upon cell stimulation. Cell migration depends on filamentous actin (F-actin) rearrangement, since pretreatment with cytochalasin B, an inhibitor of F-actin formation, prevented both DNP-HSA- and SCF-induced migration in wild-type BMMC. Immunocytochemical experiments using fluorescence-conjugated phalloidin demonstrate a reduced level of F-actin formation in DNP-HSA-stimulated BMMCs from trpm4(-/-) mice. Thus, our results suggest that TRPM4 is critically involved in migration of BMMCs by regulation of Ca(2+)-dependent actin cytoskeleton rearrangements.