EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Methicillin-resistant and -susceptible Staphylococcus aureus strains of clonal lineages ST398 and ST9 from swine carry the multidrug resistance gene cfr.
PMID 19047652
Abstract
Methicillin-resistant Staphylococcus aureus clonal lineage ST398 and methicillin-susceptible lineage ST9 strains have their main reservoir in swine but can colonize and cause infections in humans. The phenicol/lincosamide/oxazolidinone/pleuromutilin/streptogramin A multidrug resistance gene cfr was detected in isolates of both clonal lineages, rendering a spread to humans with exposure to swine farming possible.