EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Chiral recognition of alpha-phenylethylamine by sucrose-based macrocyclic receptors.
PMID 19048168
Abstract
Simple chiral aza-crown ethers based on sucrose display high enantioselectivity in complexation of phenylethylammonium chlorides.