EMAIL THIS PAGE TO A FRIEND
Pathology oncology research : POR
Gammaaminobutyric acid A receptor alpha 3 subunit is overexpressed in lung cancer.
PMID 19048400
Abstract
The identification of tumor-associated antigens, which are specifically expressed in cancer tissues, is very important for immunotherapy of lung cancer. We have combined the in silico screening and experimental verifying to identify genes that are differently expressed in cancers compared with their corresponding normal tissues. Using these methods, we have identified that GABRA3 gene was overexpressed in lung cancer and rarely expressed in other cancers. Furthermore, GABRA3 protein expression was significantly higher in the lower grade of lung cancer. It may compose functional GABA-gated channel with other subunits. This study demonstrated GABRA3 could be a potential biomarker for diagnosis of lung cancer, and GABAA receptors may play an important role in cancer differentiation.