EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
A fluorescence sensor for detection of geranyl pyrophosphate by the chemo-ensemble method.
PMID 19049370
Abstract
Hexaamide receptor 4 and coumarin phosphate 5 form an ensemble for effective sensing of geranyl pyrophosphate (GPP). Fluorescence resonance energy transfer in the 4 + 5 ensemble diminishes when 5 is replaced by GPP. Receptor 4 binds selectively with GPP over other anions, including fatty acids. The 4-GPP complex in 1:1 stoichiometry is inferred by NMR and fluorescence titrations. Receptor 4 contains a pseudotetrahedral cleft to accommodate GPP via multiple hydrogen bonds, and the two aliphatic chains exert additional hydrophobic interactions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
76532
Geranyl pyrophosphate lithium salt, ≥95.0% (TLC)
C10H20O7P2 · xLi+
19533
Geranyl pyrophosphate lithium salt, analytical standard
C10H20O7P2 · xLi+