EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Muscarinic acetylcholine receptor subtypes as agonist-dependent oncogenes.
PMID 1905013
Abstract
We have evaluated the muscarinic acetylcholine family of G protein-coupled receptors (mAChRs) for their oncogenic potential. These receptors are preferentially expressed in postmitotic cells, transducing signals specified by their endogenous agonist, the neurotransmitter acetylcholine. Cells transfected with individual human mAChR genes were morphologically indistinguishable from parental NIH 3T3 cells in the absence of agonist. In contrast, when cultures were supplemented with carbachol, a stable analog of acetylcholine, foci of transformation readily appeared in m1, m3, or m5 but not in m2 or m4 mAChRs transfectants. Receptor expression was verified by ligand binding and was similar for each transfected culture. Transformation was dose-dependent and required only low levels of receptor expression. In transformation-competent cells, agonist induced phosphatidylinositol hydrolysis, whereas in m2 or m4 transfectants, receptors were coupled to the inhibition of adenylyl cyclase. These findings demonstrate that mAChRs linked to phosphatidylinositol hydrolysis can act as conditional oncogenes when expressed in cells capable of proliferation.