EMAIL THIS PAGE TO A FRIEND
Mycopathologia
Atypical micromorphology and uncommon location of cryptococcosis: a histopathologic study using special histochemical techniques (one case report).
PMID 19052915
Abstract
Here we report an unusual case of disseminated cryptococcosis in a patient with AIDS. Although typical Cryptococcus neoformans micromorphology was observed in tongue biopsy, cervical lymph node examination revealed atypical histopathologic findings. These included pseudohyphae, chains of budding yeasts and structures resembling germ tubes. Cryptococcus neoformans infection in supraclavicular lymph nodes was also confirmed by culture. The importance of using special histochemical techniques-Mayer's mucicarmine stain for mucicarminophilic capsule and Grocott's silver stain-in the diagnosis of cryptococcosis is reinforced.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
41325 Mayer’s Mucicarmine Stain Solution, for microscopy