EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Total synthesis of (-)-norzoanthamine.
PMID 19053124
Abstract
No bones about it: (-)-Norzoanthamine, a promising candidate for an anti-osteoporotic drug, was the target of a total synthesis (see scheme). The final bisaminal formation with AcOH/H(2)O gave the DEFG ring, while the cyclization precursor was prepared by installing the remaining bisaminal unit after oxidative cleavage of the cyclopentanol moiety.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C112208
Cyclopentanol, 99%
C5H10O