EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Chloroquine-astemizole hybrids with potent in vitro and in vivo antiplasmodial activity.
PMID 19054674
Abstract
A dual activity, conjugated approach has been taken to form hybrid molecules of two known antimalarial drugs, chloroquine (CQ) and the non-sedating H1 antagonist astemizole. A variety of linkers were investigated to conjugate the two agents into one molecule. Compounds 5-8 possessed improved in vitro activity against a CQ-resistant strain of Plasmodium falciparum, and examples 7 and 8 were active in vivo in mouse models of malaria.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A2861
Astemizole, ≥98% (HPLC)
C28H31FN4O