EMAIL THIS PAGE TO A FRIEND
Cancer letters
Dickkopf-4 is frequently down-regulated and inhibits growth of colorectal cancer cells.
PMID 19059704
Abstract
Like Dickkopf-1 (DKK1), DKK4 is a target of beta-catenin/Tcf-4 in colorectal cancer. However, as a negative regulator of Wnt signalling its function in colorectal cancer cells is not well understood. We report that DKK4 is frequently down-regulated in colorectal cancer cell lines with deregulated beta-catenin/Tcf-4 and in primary colorectal cancers. Exposure of cancer cells to DKK4 strongly inhibits basal beta-catenin/Tcf-4 signalling activity, cancer cell growth and cell cycle progression. Therefore, loss of this negative feed-back loop provides Wnt factor expressing cancer cells with a growth advantage. Our data demonstrate that DKK4 is an important negative regulator of colon cancer cell growth.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
07-595
Anti-acetyl-Histone H4 (Lys12) Antibody, serum, clone, Upstate®