EMAIL THIS PAGE TO A FRIEND
Agents and actions. Supplements
A new antiinflammatory agent (EGIS-5645) without gastrointestinal side-effect.
PMID 1906236
Abstract
The compound EGIS-5645 is a potent antipyretic agent possessing analgesic and antiinflammatory properties. The drug is active in antiinflammatory models such as carrageenin oedema and adjuvant arthritis. EGIS-5645 shows practically no gastro-ulcerogenic effect. The molecule does not inhibit either prostaglandin biosynthesis or soybean lipoxygenase enzyme activity.
Related Materials