EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Structure-based pharmacophore screening for natural-product-derived PPARgamma agonists.
PMID 19067454
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S1250
(−)-α-Santonin, ≥99%
C15H18O3
53653
(−)-α-Santonin, analytical standard
C15H18O3