EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Copper(II) acetate-catalyzed addition of arylboronic acids to aromatic aldehydes.
PMID 19067580
Abstract
A novel copper-catalyzed protocol for the synthesis of carbinol derivatives has been developed. In the presence of copper(II) acetate and dppf, carbinol derivatives were prepared by the addition of arylboronic acids to aromatic aldehydes in good to excellent yields. Moreover, the rigorous exclusion of air or moisture is not required in these transformations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
517453
Copper(II) acetate, powder, 99.99% trace metals basis
C4H6CuO4
326755
Copper(II) acetate, 98%
C4H6CuO4