EMAIL THIS PAGE TO A FRIEND
Journal of inherited metabolic disease
A specific fluorogenic assay for N-acetylgalactosamine-4-sulphatase activity using immunoadsorption.
PMID 1907337
Abstract
A method combining immune capture and enzyme detection by fluorochemistry has been developed for the diagnostic assay of N-acetylgalactosamine-4-sulphatase (4-sulphatase). The procedure uses a monoclonal antibody 4-S 4.1 to immunoadsorb 4-sulphatase specifically from complex protein samples containing other sulphatases, and 4-methylumbelliferyl sulphate to detect captured 4-sulphatase. The assay provides an accurate and simple method for the diagnosis of Maroteaux-Lamy syndrome (Mucopolysaccharidosis type VI).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7133
4-Methylumbelliferyl sulfate potassium salt, sulfatase substrate
C10H7KO6S