EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Intermolecular sequential [4 + 2]-cycloaddition-aromatization reaction of aryl-substituted allenes with DMAD affording phenanthrene and naphthalene derivatives.
PMID 19082163
Abstract
An efficient entry to phenanthrene and naphthalene derivatives through intermolecular sequential [4 + 2]-cycloaddition-aromatization reactions of aryl-substituted allenes with DMAD in the absence of any catalyst was discovered. In this reaction the aromatic ring and the adjacent carbon-carbon double bond of the allene unit acted as the 1,3-diene.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D138401
Dimethyl acetylenedicarboxylate, 99%
C6H6O4