EMAIL THIS PAGE TO A FRIEND
Arthritis research & therapy
Antiphospholipid syndrome.
PMID 19090981
Abstract
Antiphospholipid syndrome is diagnosed when arterial or venous thrombosis or recurrent miscarriages occur in a person in whom laboratory tests for antiphospholipid antibodies (anticardiolipin antibodies and/or lupus anticoagulant and/or anti-beta 2-glycoprotein I) are positive. Despite the strong association between antiphospho-lipid antibodies and thrombosis, their pathogenic role in the development of thrombosis has not been fully elucidated. Novel mechanisms involving both the complement pathway and micro-particles have been described. The knowledge of these new pathogenic approaches might identify novel therapeutic targets and therefore may improve the management of these patients.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV51158
Anti-APOH antibody produced in rabbit, affinity isolated antibody