EMAIL THIS PAGE TO A FRIEND
Journal of photochemistry and photobiology. B, Biology
Photodynamic therapy with aluminum-chloro-phthalocyanine induces necrosis and vascular damage in mice tongue tumors.
PMID 19097802
Abstract
In this paper we describe the efficacy of the liposomal-AlClPc (aluminum-chloro-phthalocyanine) formulation in PDT study against Ehrlich tumor cells proliferation in immunocompetent swiss mice tongue. Experiments were conduced in sixteen tumor induced mice that were divided in three control groups: (1) tumor without treatment; (2) tumor with 100J/cm(2) laser (670nm) irradiation; and (3) tumor with AlClPc peritumoral injection; and a PDT experimental group when tumors received AlClPc injection followed by tumor irradiation. Control groups present similar macroscopically and histological patterns after treatments, while PDT treatment induced 90% of Ehrlich tumor necrosis after 24h of one single application, showing the efficacy of liposome-AlClPc (aluminum-chloro-phthalocyanine) mediated PDT on the treatment of oral cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
362530
Aluminum phthalocyanine chloride, Dye content ~85 %
C32H16AlClN8