EMAIL THIS PAGE TO A FRIEND
Sao Paulo medical journal = Revista paulista de medicina
Teriparatide (recombinant human parathyroid hormone 1-34) in postmenopausal women with osteoporosis: systematic review.
PMID 19099162
Abstract
Osteoporosis is defined as a disease characterized by low bone mass and deterioration of the bone tissue microarchitecture. Teriparatide stimulates the formation and action of osteoblasts, which are responsible for bone formation, thus promoting bone tissue increase. The aim was to assess the effectiveness and safety of teriparatide for treating postmenopausal osteoporosis. A systematic review was conducted using the Cochrane Collaboration methodology. 1) Teriparatide 20 microg or 40 microg versus placebo: there was a benefit from teriparatide, considering the following outcomes: reduction in the number of new vertebral and non-vertebral fractures, and increased whole-body, lumbar and femoral bone mineral density. 2) Teriparatide 40 microg versus alendronate 10 mg/day for 14 months: there was no statistical difference regarding the incidence of new vertebral or non-vertebral fractures, although in the group that received teriparatide there was greater bone mineral density increase in the whole body, lumbar column and femur. 3) Estrogen plus teriparatide 25 microg versus estrogen: there was a benefit, considering the following outcomes: reduction in the number of new vertebral fractures, and increased whole-body, lumbar and femoral bone mineral density after three years. When teriparatide is intermittently administered in low doses, it reduces the incidence of vertebral fractures (67%) and non-vertebral fractures (38%) and increases bone mineral density in the lumbar column and femur. There is a need for studies with longer observation in order to allow conclusions regarding the safety and duration of the therapeutic effects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7662
Nafarelin acetate salt hydrate, ≥95% (HPLC), solid
C66H83N17O13 · xC2H4O2 · yH2O