EMAIL THIS PAGE TO A FRIEND
Organic letters
ortho-Lithiophenyl isocyanide: a versatile precursor for 3H-quinazolin-4-ones and 3H-quinazolin-4-thiones.
PMID 19099490
Abstract
ortho-Lithiophenyl isocyanide has been generated from ortho-bromophenyl isocyanide and successfully employed toward the synthesis of 2-substituted phenyl isocyanides as well as 2,3-disubstituted 3H-quinazoline-4-ones and 3H-quinazolin-4-thiones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
439983
Methyl 2-isocyanatobenzoate, 97%
C9H7NO3