EMAIL THIS PAGE TO A FRIEND
Enfermedades infecciosas y microbiologia clinica
[Multifocal osteoarticular infection caused by Staphylococcus aureus: when antibiotic resistance is not the problem].
PMID 19100185
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C7020
Cefadroxil, analytical standard
C16H17N3O5S