EMAIL THIS PAGE TO A FRIEND
Journal of natural products
Cyclosporins from Mycelium sterilae MS 2929.
PMID 19102681
Abstract
The structures of two new cyclosporins were elucidated by NMR and MS methods as cyclo[-MeBmt(1)-Abu(2)-Sar(3)-MeLeu(4)-Val(5)-MeLeu(6)-Ala(7)-d-Ala(8)-MeLeu(9)-MeNva(10)-MeVal(11)-] and cyclo[-MeBmt(1)-Abu(2)-Sar(3)-MeLeu(4)-Abu(5)-MeLeu(6)-Ala(7)-d-Ala(8)-MeLeu(9)-MeLeu(10)-MeVal(11)-].
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7502
DL-Norvaline
C5H11NO2
N7627
L-Norvaline, arginase inhibitor
C5H11NO2