EMAIL THIS PAGE TO A FRIEND
Organic letters
Highly cis-selective cyclopropanations with ethyl diazoacetate using a novel Rh(I) catalyst with a chelating N-heterocyclic iminocarbene ligand.
PMID 19108673
Abstract
A structurally characterized Rh(I) iminocarbene complex (N,C)Rh(CO)Cl is activated with AgOTf to act as a highly cis-selective catalyst for the cyclopropanation of substituted styrenes and other alkenes with ethyl diazoacetate (11 examples, 10-99% yield, up to >99% cis-selectivity).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E22201
Ethyl diazoacetate, contains ≥13 wt. % dichloromethane
C4H6N2O2