EMAIL THIS PAGE TO A FRIEND
Journal of bioscience and bioengineering
Isolation of industrial strains of Aspergillus oryzae lacking ferrichrysin by disruption of the dffA gene.
PMID 19111645
Abstract
Based on studies using laboratory strains, the efficiency of gene disruption in Aspergillus oryzae, commonly known as koji mold, is low; thus, gene disruption has rarely been applied to the breeding of koji mold. To evaluate the efficiency of gene disruption in industrial strains of A. oryzae, we produced ferrichrysin biosynthesis gene (dffA) disruptants using three different industrial strains as hosts. PCR analysis of 438 pyrithiamine-resistant transformants showed dffA gene disruption efficiency of 42.9%-64.1%, which is much higher than previously reported. Analysis of the physiological characteristics of the disruptants indicated that dffA gene disruption results in hypersensitivity to hydrogen peroxide. To investigate the industrial characteristics of dffA gene disruptants, two strains were used to make rice koji and their properties were compared to those of the host strains. No differences were found between the dffA gene disruptants and the host strains, except that the disruptants did not produce ferrichrysin. Thus, this gene disruption technique is much more effective than conventional mutagenesis for A. oryzae breeding.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P0256
Pyrithiamine hydrobromide, ~95%, crystalline
C14H18N4O · 2HBr